ENA388700730 Enamine

1-(4-methylpyridin-3-yl)ethan-1-amine

  • MDL number MFCD13189155
  • Cas Number : 1060805-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124807
Availability details for
Quantity
  • Availability
  • Add to Cart