ENA388700731 Enamine

(1-cyclobutylethyl)(methyl)amine

  • MDL number MFCD21117757
  • Cas Number : 1427379-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124808
Availability details for
Quantity
  • Availability
  • Add to Cart