ENA388700735 Enamine

1-(2-methylphenyl)-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD17328144
  • Cas Number : 1273930-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124666
Availability details for
Quantity
  • Availability
  • Add to Cart