ENA388700749 Enamine

2-chloro-4-(methylsulfanyl)benzonitrile

  • MDL number MFCD17256842
  • Cas Number : 1002557-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124714

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124714
Availability details for
Quantity
  • Availability
  • Add to Cart