ENA388700751 Enamine

2-methyl-1-[4-(3-methylphenyl)piperazin-1-yl]propan-2-amine

  • MDL number MFCD13327244
  • Cas Number : 1305785-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124717
Availability details for
Quantity
  • Availability
  • Add to Cart