ENA388700754 Enamine

4-[(4-chloro-2-methylphenyl)amino]benzonitrile

  • MDL number MFCD11124643
  • Cas Number : 1039838-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124718
Availability details for
Quantity
  • Availability
  • Add to Cart