ENA388700755 Enamine

4-methoxy-3-(1-methyl-1H-pyrazol-3-yl)benzoic acid

  • MDL number MFCD23144247
  • Cas Number : 1427379-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124719
Availability details for
Quantity
  • Availability
  • Add to Cart