ENA388700756 Enamine

{1-[(4-chlorophenyl)methyl]cyclopropyl}methanol

  • MDL number MFCD21671059
  • Cas Number : 1427379-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124720
Availability details for
Quantity
  • Availability
  • Add to Cart