ENA388700763 Enamine

1-[1-(pyrimidin-2-yl)piperidin-3-yl]methanamine

  • MDL number MFCD08669759
  • Cas Number : 912761-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124654
Availability details for
Quantity
  • Availability
  • Add to Cart