ENA388700766 Enamine

N-[2-(morpholin-4-yl)ethyl]-3-(trifluoromethyl)aniline dihydrochloride

  • MDL number MFCD23144286
  • Cas Number : 1427379-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124884
Availability details for
Quantity
  • Availability
  • Add to Cart