ENA388700767 Enamine

4-amino-N-(3-methoxypyrazin-2-yl)benzene-1-sulfonamide

  • MDL number MFCD00437754
  • Cas Number : 152-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124886
Availability details for
Quantity
  • Availability
  • Add to Cart