ENA388700769 Enamine

4-(2,3-dihydro-1-benzofuran-5-yl)-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD11180042
  • Cas Number : 1094394-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124891
Availability details for
Quantity
  • Availability
  • Add to Cart