ENA388700778 Enamine

3-bromo-5-methoxypyridin-2-amine

  • MDL number MFCD12025479
  • Cas Number : 1379325-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124677
Availability details for
Quantity
  • Availability
  • Add to Cart