ENA388700784 Enamine

1-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-(morpholin-4-yl)propan-1-one hydrochloride

  • MDL number MFCD00097056
  • Cas Number : 22310-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124736
Availability details for
Quantity
  • Availability
  • Add to Cart