ENA388700790 Enamine

2-(1-chloroethyl)-5-methyl-1,3-oxazole

  • MDL number MFCD20231587
  • Cas Number : 1334485-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124974
Availability details for
Quantity
  • Availability
  • Add to Cart