ENA388700791 Enamine

3-ethyl-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08700651
  • Cas Number : 113100-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124975

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124975
Availability details for
Quantity
  • Availability
  • Add to Cart