ENA388700792 Enamine

4-(chloromethyl)-5-(propan-2-yl)-1,3-oxazole

  • MDL number MFCD21809055
  • Cas Number : 1427379-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124976
Availability details for
Quantity
  • Availability
  • Add to Cart