ENA388700796 Enamine

6-bromo-1-ethyl-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD21194035
  • Cas Number : 24810-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124982
Availability details for
Quantity
  • Availability
  • Add to Cart