ENA388700800 Enamine

6-amino-7-[(3-hydroxybutyl)amino]-3,4-dihydroquinazolin-4-one

  • MDL number MFCD20123782
  • Cas Number : 1427379-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124987
Availability details for
Quantity
  • Availability
  • Add to Cart