ENA388700801 Enamine

4-(dimethylamino)pyridine-3-carbaldehyde

  • MDL number MFCD18384613
  • Cas Number : 90006-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-68
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124988
Availability details for
Quantity
  • Availability
  • Add to Cart