ENA388700802 Enamine

2-[1-(pyridin-4-yl)piperidin-4-yl]ethan-1-amine

  • MDL number MFCD16806706
  • Cas Number : 625438-12-2
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[1-(4-pyridinyl)-4-piperidinyl]ethanamine
Inchi Code 1S/C12H19N3/c13-6-1-11-4-9-15(10-5-11)12-2-7-14-8-3-12/h2-3,7-8,11H,1,4-6,9-10,13H2
Inchi Key UMDLIABIZYDUHP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124992
Availability details for
Quantity
  • Availability
  • Add to Cart