ENA388700807 Enamine

4-[3-(trifluoromethoxy)phenyl]-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD23144295
  • Cas Number : 1427380-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125004
Availability details for
Quantity
  • Availability
  • Add to Cart