ENA388700808 Enamine

3-phenyl-2,5-dihydro-1H-pyrrole hydrochloride

  • MDL number MFCD23144296
  • Cas Number : 97382-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125005
Availability details for
Quantity
  • Availability
  • Add to Cart