ENA388700809 Enamine

3-(2-methylphenyl)-2,5-dihydro-1H-pyrrole hydrochloride

  • MDL number MFCD23144297
  • Cas Number : 1427378-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125006
Availability details for
Quantity
  • Availability
  • Add to Cart