ENA388700811 Enamine

1-(1-benzofuran-2-yl)-3-(morpholin-4-yl)propan-1-one

  • MDL number MFCD01461070
  • Cas Number : 124617-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124738
Availability details for
Quantity
  • Availability
  • Add to Cart