ENA388700813 Enamine

1-(1-benzofuran-2-yl)-3-(dimethylamino)propan-1-one hydrochloride

  • MDL number MFCD00438735
  • Cas Number : 15817-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124739

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124739
Availability details for
Quantity
  • Availability
  • Add to Cart