ENA388700818 Enamine

2-(4-amino-2,3-dihydro-1H-isoindol-2-yl)propan-1-ol

  • MDL number MFCD10007261
  • Cas Number : 1017462-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125044
Availability details for
Quantity
  • Availability
  • Add to Cart