ENA388700822 Enamine

1-(bromomethyl)-3-methylcyclohexane

  • MDL number MFCD21296516
  • Cas Number : 1427380-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125062
Availability details for
Quantity
  • Availability
  • Add to Cart