ENA388700827 Enamine

3-(morpholin-4-yl)-1-(naphthalen-2-yl)propan-1-one hydrochloride

  • MDL number MFCD00111282
  • Cas Number : 674284-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124746

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124746
Availability details for
Quantity
  • Availability
  • Add to Cart