ENA388700829 Enamine

3-[4-(propan-2-yloxy)phenyl]propanal

  • MDL number MFCD20336805
  • Cas Number : 683746-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125081
Availability details for
Quantity
  • Availability
  • Add to Cart