ENA388700838 Enamine

2-(4,5-dihydro-1H-imidazol-2-yl)acetonitrile

  • MDL number MFCD15833258
  • Cas Number : 23539-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...86
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124749

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124749
Availability details for
Quantity
  • Availability
  • Add to Cart