ENA388700840 Enamine

2-chloro-6-sulfanylbenzonitrile

  • MDL number MFCD00191230
  • Cas Number : 72371-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125110
Availability details for
Quantity
  • Availability
  • Add to Cart