ENA388700841 Enamine

4-amino-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD00233927
  • Cas Number : 1003-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124649

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124649
pictograms GHS01,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart