ENA388700844 Enamine

N-(2,3-dihydro-1,4-benzodioxin-6-yl)butanamide

  • MDL number MFCD00248184
  • Cas Number : 423151-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-125155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125155
Availability details for
Quantity
  • Availability
  • Add to Cart