ENA388700847 Enamine

5-(3-acetylphenoxymethyl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD16385637
  • Cas Number : 1281554-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124765

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124765
Availability details for
Quantity
  • Availability
  • Add to Cart