ENA388700855 Enamine

4-{4-[2-(4-chlorophenoxy)ethyl]piperazin-1-yl}-3-nitrobenzoic acid

  • MDL number MFCD23144262
  • Cas Number : 1427379-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-124775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124775
Availability details for
Quantity
  • Availability
  • Add to Cart