ENA388700856 Enamine

2-(pentafluoroethyl)-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD11619463
  • Cas Number : 1094485-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124778

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124778
Availability details for
Quantity
  • Availability
  • Add to Cart