ENA388700875 Enamine

4-methoxypyrrolidine-2-carboxylic acid

  • MDL number MFCD19596805

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31040

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31040
Availability details for
Quantity
  • Availability
  • Add to Cart