ENA388700881 Enamine

4-bromo-1-N-ethylbenzene-1,2-diamine

  • MDL number MFCD10689144
  • Cas Number : 334829-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45068
Availability details for
Quantity
  • Availability
  • Add to Cart