ENA388700891 Enamine

3-(chlorosulfonyl)-2,6-dimethoxybenzoic acid

  • MDL number MFCD07774204
  • Cas Number : 874841-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56415

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56415
Availability details for
Quantity
  • Availability
  • Add to Cart