ENA388700893 Enamine

1-(3-methoxy-4-methylphenyl)methanamine

  • MDL number MFCD09809087
  • Cas Number : 247570-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58644
Availability details for
Quantity
  • Availability
  • Add to Cart