ENA388700896 Enamine

1-cyclopropylprop-2-en-1-one

  • MDL number MFCD18975090
  • Cas Number : 59819-62-4
  • Molecular Weight : 96.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropylprop-2-en-1-one
Inchi Code 1S/C6H8O/c1-2-6(7)5-3-4-5/h2,5H,1,3-4H2
Inchi Key TWFKEGONKTUVSX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60337
Availability details for
Quantity
  • Availability
  • Add to Cart