ENA388700907 Enamine

3-[3-(dimethylamino)propoxy]aniline

  • MDL number MFCD09746237
  • Cas Number : 1135-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74750

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74750
Availability details for
Quantity
  • Availability
  • Add to Cart