ENA388700917 Enamine

oxepan-4-one

  • MDL number MFCD20484206
  • Cas Number : 62643-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85698
Availability details for
Quantity
  • Availability
  • Add to Cart