ENA388700934 Enamine

2-fluoro-2-methylpropanoic acid

  • MDL number MFCD03452788
  • Cas Number : 63812-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-95556

Safety Information

signalword Danger
pcodes P260,P264,P270,P271,P280,P284,P301+P310,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P320,P330,P361,P363,P403+P233,P405,P501
hcodes C,T
MSDS https://www.enaminestore.com/catalog/EN300-95556
pictograms GHS05,GHS06
hstatements H301,H311,H314,H330
Availability details for
Quantity
  • Availability
  • Add to Cart