ENA388700936 Enamine

3-amino-2,3-dimethylbutan-2-ol

  • MDL number MFCD19203546
  • Cas Number : 89585-13-7
  • Molecular Weight : 117.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-2,3-dimethyl-2-butanol
Inchi Code 1S/C6H15NO/c1-5(2,7)6(3,4)8/h8H,7H2,1-4H3
Inchi Key OZSQDNCDEHVGBC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-97040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97040
Availability details for
Quantity
  • Availability
  • Add to Cart