ENA390346758 Enamine

octahydro-1H-indole-6-sulfonic acid

  • MDL number MFCD23701777
  • Cas Number : 1432679-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116358
Availability details for
Quantity
  • Availability
  • Add to Cart