ENA390346759 Enamine

hexahydro-2H-cyclopenta[b][1,4]dithiin-5-one

  • MDL number MFCD23701778
  • Cas Number : 1432679-71-4
  • Molecular Weight : 174.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name hexahydro-5H-cyclopenta[b][1,4]dithiin-5-one
Inchi Code 1S/C7H10OS2/c8-5-1-2-6-7(5)10-4-3-9-6/h6-7H,1-4H2
Inchi Key RTWLIAGBLSHCBR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-118158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118158
Availability details for
Quantity
  • Availability
  • Add to Cart