ENA390346760 Enamine

2-methyl-4-{4H,5H,6H,7H-thieno[3,2-c]pyridin-2-yl}-1,3-thiazole

  • MDL number MFCD23701779
  • Cas Number : 1432682-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118478

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118478
Availability details for
Quantity
  • Availability
  • Add to Cart