ENA390346761 Enamine

[2-(2,2-difluoroethoxy)pyridin-4-yl]methanamine

  • MDL number MFCD23701780
  • Cas Number : 1432680-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118957

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-118957
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart